missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, silver salt, 99%

Catalog No. AC419340250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419340250 25 g Glass bottle
AC419340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC419340250 Supplier Thermo Scientific Chemicals Supplier No. 419340250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2386-52-9 | CH3AgO3S | 202.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2386-52-9
Molecular Formula CH3AgO3S
Molecular Weight (g/mol) 202.97
MDL Number MFCD00064795
InChI Key MLKQJVFHEUORBO-UHFFFAOYSA-M
Synonym silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
PubChem CID 159658
IUPAC Name silver;methanesulfonate
SMILES CS(=O)(=O)[O-].[Ag+]

Specifications

Melting Point 252.0°C to 256.0°C
Color Beige to White or Gray
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula CH3SO3Ag
Beilstein 04,4
Solubility Information Solubility in water: soluble.
Formula Weight 202.97
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methanesulfonic acid, silver salt, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a po
EINECSNumber 219-199-2
RTECSNumber PB2685000
TSCA TSCA

RUO – Research Use Only