missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacrolein, 90%, stabilized, AcroSeal™, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Density | 0.8470g/mL |
| Boiling Point | 69.0°C |
| Flash Point | −15°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 88% min. (GC) |
| Packaging | AcroSeal™ Glass bottle |
| Linear Formula | H2C=C(CH3)CHO |
| Refractive Index | 1.4145 |
| Quantity | 100mL |
| Beilstein | 01,III,2981 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Fatal if inhaled. Toxic in contact with skin. Toxic if swallowed. Causes serious eye damage. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 201-150-1 |
RUO – Research Use Only