missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metaphosphoric acid, ACS reagent, stabilized
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Packaging | Glass Bottle |
| Quantity | 100 g |
| Linear Formula | (HPO3)n |
| Solubility Information | Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol |
| Grade | ACS Reagent |
| Chemical Name or Material | Metaphosphoric acid |
RUO – Research Use Only