missing translation for 'onlineSavingsMsg'
Learn More

Metaphosphoric Acid, 3% w/v, Reagents

Catalog No. 01334763
Change view
Click to view available options
Quantity:
500 mL
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334763 10 L
01334762 20 L
01334761 4 L
01334760 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01334763 Supplier Labchem Inc Supplier No. CM10350010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Metaphosphoric Acid, 3% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Phosphenic Acid
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.019
Quantity 10 L
Physical Form Liquid
Percent Purity 3%
Grade Reagent
Chemical Name or Material Metaphosphoric Acid, 3% w/v