missing translation for 'onlineSavingsMsg'
Learn More

Metanilic acid, 98%, Thermo Scientific™

Numéro de catalogue. AC413712500
Modifier l'affichage
Click to view available options
Quantité:
100g
250g
1kg
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC413712500 250g Glass bottle
AC413711000 100g Glass bottle
AC413710010 1kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC413712500 Fournisseur Thermo Scientific Chemicals Code fournisseur 413712500
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 121-47-1
Formule moléculaire C6H7NO3S
Poids moléculaire (g/mol) 173.19
Numéro MDL MFCD00065345
Clé InChI ZAJAQTYSTDTMCU-UHFFFAOYSA-N
Synonyme metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
PubChem CID 8474
ChEBI CHEBI:27764
Nom de l’IUPAC 3-aminobenzenesulfonic acid
SOURIRES C1=CC(=CC(=C1)S(=O)(=O)O)N

Spécifications

Nom ou substance chimique Metanilic acid
Nom Remarque 98%
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98%
Emballage Glass bottle
Formule linéaire NH2C6H4SO3H
Quantité 250g
Indice de Merck 15, 5998
Renseignements sur la solubilité Solubility in water: slightly soluble
Poids de formule 173.19
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-473-6
RTECSNumber OY2300000
TSCA TSCA

RUO – Research Use Only