Learn More
Beckman Coulter Met One Particle Counter Printer Paper
Catalog No. 1899913C
Encompass
Change view
Click to view available options
Product Type:
Printer Paper
Catalog No. Product Type
1899913C Printer Paper
1 options
Catalog No. 1899913C Supplier Beckman Coulter Supplier No. 460519
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Product Type Printer Paper
Quantity Each
Width (English) 2 in.
Width (Metric) 5.1cm
For Use With (Equipment) Thermal printer