missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Oxide, Approx. 98%, Spectrum™ Chemical

Catalog No. 18604336
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604336 2.5 L Amber Glass Bottle
18604334 100 mL Amber Glass Bottle
18604335 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604336 Supplier Spectrum Chemical Mfg Cor Supplier No. M24912.5LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M2491, 141-79-7, C6H10O

Spectrum™ Chemical Mesityl Oxide is a colorless compound with a strong peppermint scent. It is known to be a volatile liquid. Mesityl oxide is produced under the same conditions of isophorone. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(C)=CC(C)=O

Specifications

CAS Max % 100%
Boiling Point 130°C (266°F)
Packaging Amber Glass Bottle
Quantity 2.5 L
UN Number UN1229
Percent Purity ∼98%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material Mesityl Oxide