missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesityl Fluoride 98.0+%, TCI America™
$119.36 - $344.61
Chemical Identifiers
| CAS | 392-69-8 |
|---|---|
| Molecular Formula | C9H11F |
| Molecular Weight (g/mol) | 138.19 |
| MDL Number | MFCD00052365 |
| InChI Key | ZLGPNBBJPOBSLY-UHFFFAOYSA-N |
| Synonym | fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 |
| PubChem CID | 136234 |
| IUPAC Name | 2-fluoro-1,3,5-trimethylbenzene |
| SMILES | CC1=CC(C)=C(F)C(C)=C1 |
Chemical Identifiers
| 392-69-8 | |
| 138.19 | |
| ZLGPNBBJPOBSLY-UHFFFAOYSA-N | |
| 136234 | |
| CC1=CC(C)=C(F)C(C)=C1 |
| C9H11F | |
| MFCD00052365 | |
| fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 | |
| 2-fluoro-1,3,5-trimethylbenzene |
Specifications
| 392-69-8 | |
| 165°C | |
| MFCD00052365 | |
| fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 | |
| CC1=CC(C)=C(F)C(C)=C1 | |
| 138.19 | |
| 138.19 | |
| Liquid |
| Yellow | |
| C9H11F | |
| 1 g | |
| ZLGPNBBJPOBSLY-UHFFFAOYSA-N | |
| 2-fluoro-1,3,5-trimethylbenzene | |
| 136234 | |
| ≥98.0% (GC) | |
| Mesityl Fluoride |
Safety and Handling
TSCA : No