accessibility menu, dialog, popup

UserName

CAS RN 392-69-8

IUPAC Name: 2-fluoro-1,3,5-trimethylbenzene
Synonyms: 2-fluoro-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 138.19
Molecular Formula: C9H11F
InChi Key: ZLGPNBBJPOBSLY-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(F)C(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)