missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesityl Fluoride 98.0+%, TCI America™
Supplier: TCI America M12015G
Specifications
| Mesityl Fluoride | |
| Yellow | |
| C9H11F | |
| 5 g | |
| ZLGPNBBJPOBSLY-UHFFFAOYSA-N | |
| 2-fluoro-1,3,5-trimethylbenzene | |
| 136234 | |
| ≥98.0% (GC) |
| 392-69-8 | |
| 165°C | |
| MFCD00052365 | |
| fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 | |
| CC1=CC(C)=C(F)C(C)=C1 | |
| 138.19 | |
| 138.19 | |
| Liquid |
Chemical Identifiers
| 392-69-8 | |
| 138.19 | |
| ZLGPNBBJPOBSLY-UHFFFAOYSA-N | |
| 136234 | |
| CC1=CC(C)=C(F)C(C)=C1 |
| C9H11F | |
| MFCD00052365 | |
| fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939 | |
| 2-fluoro-1,3,5-trimethylbenzene |
Safety and Handling
TSCA : No