missing translation for 'onlineSavingsMsg'
Learn More

Mesalamine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111015757
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015757 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015757 Supplier Merck Emd Millipore Supplier No. PHR10601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mesalamine is a drug, which is structurally similar to salicylates. It exhibits anti-inflammatory activity and is widely used as a medication for the treatment of inflammatory bowel disease, including ulcerative proctitis and Crohn′s disease.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 89-57-6
Physical Form Neat
Linear Formula H2NC6H3-2-(OH)CO2H
MDL Number MFCD00007877
Melting Point 275°C to 280°C (decomposition)
Synonym 5-Aminosalicylic acid; 5-AS; 5-Amino-2-hydroxybenzoic acid; Mesalamine
Quantity 1 g
Formula Weight 153.14
Grade Certified Reference Material