missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES sodium salt, 99%

Catalog No. AC397350250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397350250 25 g Plastic bottle
AC397351000 100 g Plastic bottle
AC397355000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC397350250 Supplier Thermo Scientific Chemicals Supplier No. 397350250
Only null left

CAS: 71119-23-8 | C6H12NNaO4S | 217.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 71119-23-8
Molecular Formula C6H12NNaO4S
Molecular Weight (g/mol) 217.215
MDL Number MFCD00065473
InChI Key IRHWMYKYLWNHTL-UHFFFAOYSA-M
PubChem CID 23673676
ChEBI CHEBI:62955
IUPAC Name sodium;2-morpholin-4-ylethanesulfonate
SMILES C1COCCN1CCS(=O)(=O)[O-].[Na+]

Specifications

Absorbance 0.03 max. (10% in water) 1 cm cell vs water at 280nm, 0.05 max. (10% in water) 1 cm cell vs water at 260nm
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Information Solubility in water: 0.5g/L.
Formula Weight 217.22
Percent Purity ≥98.5% (after ion exchange)
Chemical Name or Material MES sodium salt
Show More Show Less

RUO – Research Use Only