missing translation for 'onlineSavingsMsg'
Learn More

MES, Monohydrate, cGMP Manufactured, 99.5%, Spectrum™ Chemical

Catalog No. p-7157779
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604325 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604325 Supplier Spectrum Chemical Mfg Cor Supplier No. M28481KGBL
Only null left

M2848, 145224-94-8, C6H13NO4S·H2O

Spectrum™ Chemical MES, Monohydrate, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 213.25
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
IUPAC Name 4-(2-sulfonatoethyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CC[NH+]1CCOCC1

Specifications

CAS Max % 100%
pH 2.5 to 4.0
Physical Form Powder
Quantity 1 kg
Packaging Poly Bottle
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material 2-(N-Morpholino)Ethanesulfonic Acid