missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry

CAS: 145224-94-8 | C6H15NO5S | 195.23 g/mol

$361.71 - $4848.09

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 195.23
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-(N-Morpholino)ethanesulfonic acid monohydrate
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC327761000
SDS View Documents
Thermo Scientific Chemicals
327761000
100 g Plastic bottle
Each for $361.71
Only null left
 
AC327765000
SDS View Documents
Thermo Scientific Chemicals
327765000
500 g Plastic bottle
Each for $1,275.34
Only null left
 
AC327760025
SDS View Documents
Thermo Scientific Chemicals
327760025
2.5 kg Plastic bottle
Each for $4,848.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

145224-94-8
195.23
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
C1COCCN1CCS(=O)(=O)O.O
C6H15NO5S
MFCD00149409
2-(N-Morpholino)ethanesulfonic acid monohydrate
2-morpholin-4-ylethanesulfonic acid;hydrate

Specifications

(0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
>300.0°C
Orange-Yellow to White
Crystals, Flakes or Powder
99+%
C6H15NO5S
Plastic bottle
Solubility in water: soluble.
C1COCCN1CCS(=O)(=O)O.O
195.23
213.25
Biochemical
(0.1M aq. soln.)
145224-94-8
2.5 to 4.0 (1%, H2O, 25°C)
100 g
Authentic
MFCD00149409
2-(N-Morpholino)ethanesulfonic acid monohydrate
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
16218417
≥99%
MES monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS
Documents

Documents

RUO – Research Use Only