missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry

Catalog No. AC327765000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327765000 500 g Plastic bottle
AC327761000 100 g Plastic bottle
AC327760025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC327765000 Supplier Thermo Scientific Chemicals Supplier No. 327765000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 145224-94-8 | C6H15NO5S | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-(N-Morpholino)ethanesulfonic acid monohydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Absorbance (0.1M in H2O) 1 cm cell, (0.1M in H2O) 1 cm cell at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point >300.0°C
Color Orange-Yellow to White
pH 2.5 to 4.0 (1%, H2O, 25°C)
Physical Form Crystals, Flakes or Powder
Quantity 500 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Information Solubility in water: soluble.
Formula Weight 213.25
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material MES monohydrate
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

RUO – Research Use Only