Learn More
Thermo Scientific Chemicals MES monohydrate, 98%
CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol
$88.63 - $826.96
Chemical Identifiers
| CAS | 145224-94-8 |
|---|---|
| Molecular Formula | C6H15NO5S |
| Molecular Weight (g/mol) | 213.248 |
| MDL Number | MFCD00149409 |
| InChI Key | MIIIXQJBDGSIKL-UHFFFAOYSA-N |
| Synonym | 4-Morpholineethanesulfonic acid monohydrate, 2-(4-Morpholinyl)ethanesulfonic acid monohydrate |
| PubChem CID | 16218417 |
| IUPAC Name | 2-morpholin-4-ylethanesulfonic acid;hydrate |
| SMILES | C1COCCN1CCS(=O)(=O)O.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1610414
|
Thermo Scientific Chemicals
A1610414 |
25 g |
N/A
|
|
|||||
|
AAA1610422
|
Thermo Scientific Chemicals
A1610422 |
100 g |
N/A
|
|
|||||
|
AAA1610436
|
Thermo Scientific Chemicals
A1610436 |
500 g |
N/A
|
|
|||||
Description
MES monohydrate is a biological buffer often referred to as Goodâs buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMES monohydrate is a biological buffer often referred to as Goodâs buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 145224-94-8 | |
| 213.248 | |
| MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
| 16218417 | |
| C1COCCN1CCS(=O)(=O)O.O |
| C6H15NO5S | |
| MFCD00149409 | |
| 4-Morpholineethanesulfonic acid monohydrate, 2-(4-Morpholinyl)ethanesulfonic acid monohydrate | |
| 2-morpholin-4-ylethanesulfonic acid;hydrate |
Specifications
| ∼308°C (decomposition) | |
| 25 g | |
| C6H15NO5S | |
| 141319 | |
| 4-Morpholineethanesulfonic acid monohydrate, 2-(4-Morpholinyl)ethanesulfonic acid monohydrate | |
| MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
| 2-morpholin-4-ylethanesulfonic acid;hydrate | |
| 16218417 | |
| 98% |
| 145224-94-8 | |
| 98% | |
| MFCD00149409 | |
| 14,5902 | |
| Soluble in water. | |
| C1COCCN1CCS(=O)(=O)O.O | |
| 213.248 | |
| 213.26 (195.24 Anhydrous) | |
| MES monohydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 224-632-3
RTECSNumber : QE3479500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only