Learn More
Thermo Scientific Chemicals MES, 99+%, <1% water
CAS: 4432-31-9 | C6H13NO4S | 195.233 g/mol
$219.40 - $2,309.82
Chemical Identifiers
| CAS | 4432-31-9 |
|---|---|
| Molecular Formula | C6H13NO4S |
| Molecular Weight (g/mol) | 195.233 |
| MDL Number | MFCD00006181 |
| InChI Key | SXGZJKUKBWWHRA-UHFFFAOYSA-N |
| Synonym | 2-(N-Morpholino)ethanesulfonic acid |
| PubChem CID | 78165 |
| ChEBI | CHEBI:39005 |
| IUPAC Name | 2-morpholin-4-ylethanesulfonic acid |
| SMILES | C1COCCN1CCS(=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC274140500
|
Thermo Scientific Chemicals
274140500 |
50 g | Glass bottle |
N/A
|
|
||||
|
AC274142500
|
Thermo Scientific Chemicals
274142500 |
250 g | Glass bottle |
N/A
|
|
||||
|
AC274140010
|
Thermo Scientific Chemicals
274140010 |
1 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
| 4432-31-9 | |
| 195.233 | |
| SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
| 78165 | |
| 2-morpholin-4-ylethanesulfonic acid |
| C6H13NO4S | |
| MFCD00006181 | |
| 2-(N-Morpholino)ethanesulfonic acid | |
| CHEBI:39005 | |
| C1COCCN1CCS(=O)(=O)O |
Specifications
| >300.0°C | |
| White | |
| 50 g | |
| Authentic | |
| MFCD00006181 | |
| 15, 5972 | |
| Solubility in water: 10g/100mL. | |
| C1COCCN1CCS(=O)(=O)O | |
| 195.233 | |
| CHEBI:39005 | |
| ≥99% |
| 4432-31-9 | |
| Powder | |
| 99+% | |
| C6H13NO4S | |
| Glass bottle | |
| 2-(N-Morpholino)ethanesulfonic acid | |
| SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
| 2-morpholin-4-ylethanesulfonic acid | |
| 78165 | |
| 195.24 | |
| MES |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Use only outdoors or in a well-ventilated area.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only