Learn More
Thermo Scientific Chemicals MES, 99+%, <1% water
CAS: 4432-31-9 | C6H13NO4S | 195.233 g/mol
Supplier: Thermo Scientific Chemicals 274142500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
| MES | |
| 4432-31-9 | |
| Powder | |
| 99+% | |
| C6H13NO4S | |
| Glass bottle | |
| 2-(N-Morpholino)ethanesulfonic acid | |
| SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
| 2-morpholin-4-ylethanesulfonic acid | |
| 78165 | |
| 195.24 |
| >300.0°C | |
| White | |
| 250 g | |
| Authentic | |
| MFCD00006181 | |
| 15, 5972 | |
| Solubility in water: 10g/100mL. | |
| C1COCCN1CCS(=O)(=O)O | |
| 195.233 | |
| CHEBI:39005 | |
| ≥99% |
Chemical Identifiers
| 4432-31-9 | |
| 195.233 | |
| SXGZJKUKBWWHRA-UHFFFAOYSA-N | |
| 78165 | |
| 2-morpholin-4-ylethanesulfonic acid |
| C6H13NO4S | |
| MFCD00006181 | |
| 2-(N-Morpholino)ethanesulfonic acid | |
| CHEBI:39005 | |
| C1COCCN1CCS(=O)(=O)O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Use only outdoors or in a well-ventilated area.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only