missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 6.5

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

$299.44 - $454.38

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61587AK
View Documents
Thermo Scientific Chemicals
J61587AK
250 mL
Each for $299.44
Only null left
 
AAJ61587AP
View Documents
Thermo Scientific Chemicals
J61587AP
500 mL
Each for $454.38
Only null left
 
Description

Description

MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

145224-94-8
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
C6H15NO5S
MFCD00283848
16218417
C1COCCN1CCS(=O)(=O)O.O

Specifications

145224-94-8
6.5
250 mL
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid;hydrate
16218417
1.0 M
Liquid
C6H15NO5S
Miscible with water.
C1COCCN1CCS(=O)(=O)O.O
213.248
MES buffer soln.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only