missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 5.5

Catalog No. AAJ63257AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63257AK 250 mL
AAJ63257AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63257AK Supplier Thermo Scientific Chemicals Supplier No. J63257AK

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Color Colorless
Concentration 0.2 M
pH 5.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible.
Chemical Name or Material MES buffer soln.
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only