missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

Catalog No. CLHG42Y
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLHG42Y 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLHG42Y Supplier SPEX CertiPrep Group LLC Supplier No. CLHG42Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hg1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7439-97-6 , 7697-37-2 , 7732-18-5
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 10.0, 89.9
CAS Max % 0.1, 10.0, 89.9
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Solvent or Matrix 10% HNO3
Quantity 125 mL
Grade ICP/ICP-MS
Chemical Name or Material Mercury Standard
Show More Show Less
Hazard Category Causes skin irritation, serious eye irritation
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only