Learn More
Mercury Standard For ICP, 1000μg/mL in 10% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLHG42X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Hg | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Assurance Grade Mercury Standard | |
| 7439-97-6 , 7697-37-2 , 7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.0,5.0,94.99 | |
| MFCD00011035 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| QSHDDOUJBYECFT-UHFFFAOYSA-N | |
| 10% HNO3 | |
| 200.59 | |
| 23931 | |
| Assurance |
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 0.0,5.0,94.99 | |
| Hg | |
| AA, ICP | |
| Certified Reference Material | |
| [Hg] | |
| mercury | |
| 500 mL | |
| CHEBI:16170 |
Chemical Identifiers
| 7439-97-6 | |
| 200.59 | |
| QSHDDOUJBYECFT-UHFFFAOYSA-N | |
| CHEBI:16170 | |
| [Hg] |
| Hg | |
| MFCD00011035 | |
| 23931 | |
| mercury |
Safety and Handling
Causes severe skin burns and eye damage May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful to aquatic life with long lasting effects.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only