missing translation for 'onlineSavingsMsg'
Learn More

Mercury Standard, 20 ppm Hg, Ricca Chemical

Catalog No. RTCLPHG100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RTCLPHG100 100 mL Natural LDPE Bottle
RTCLPHG500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. RTCLPHG100 Supplier Ricca Chemical Company Supplier No. RTCLPHG100
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 7439-97-6 , 7697-37-2 , 7732-18-5
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 91.0, 6.84, 0.0016
CAS Max % 95.0, 7.12, 0.0016
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 0.02mg Hg, 20ppm Hg
Chemical Name or Material Mercury Standard
ShelfLife 12 months