missing translation for 'onlineSavingsMsg'
Learn More

Mercury Organometallic Standards, SPEX CertiPrep™

Catalog No. ORGHG82Z
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ORGHG82Z 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ORGHG82Z Supplier SPEX CertiPrep Group LLC Supplier No. ORGHG82Z

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hg1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Mercury Oil Standard
Packaging Triple-leached LDPE bottle
CAS 7439-97-6, 64742-65-0
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Physical Form Fluid
Molecular Formula Hg
MDL Number MFCD00011035
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
SMILES [Hg]
Solvent or Matrix Base Oil 75
IUPAC Name mercury
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 200.59
Quantity 50 g
PubChem CID 23931
ChEBI CHEBI:16170
Show More Show Less

Product intended for laboratory use only