missing translation for 'onlineSavingsMsg'
Learn More

Mercury ICP-MS Standard, 10,000 ppm Hg in 10% HNO3, Ricca Chemical

Catalog No. MSHG10KN100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
MSHG10KN100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. MSHG10KN100 Supplier Ricca Chemical Company Supplier No. MSHG10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 7439-97-6 , 7697-37-2 , 7732-18-5
Molecular Formula Hg
Molecular Weight (g/mol) 200.592
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Hg
CAS Min % 82.0, 15.37, 0.82
CAS Max % 85.0, 15.99, 0.85
Equivalence 1 mL = 10 mg Hg
Chemical Name or Material Mercury ICP-MS Standard