missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury GFAA Cal/Spike Standard - 100 ppm Hg in 5% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 91.0, 6.8, 0.01 |
| CAS Max % | 95.0, 7.08, 0.01 |
| Concentration or Composition (by Analyte or Components) | 100ppm Hg in 5% HNO3 |
| Chemical Name or Material | Mercury GFAA Cal / Spike Standard |
Safety and Handling
| ShelfLife | 12 months |