missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Hg), LabChem™
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 89.88, 9.94, 0.18 |
| Identification | Passes Test |
| Viscosity | 0.99cSt |
| Density | 1.05g/mL |
| Physical Form | Liquid |
| Concentration | 1000 ppm ±10 ppm (1mL = 1mg Hg) |
| Linear Formula | Hg(NO3)2·H2O |
| UN Number | UN3264 |
| Color | Colorless |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause damage to organs (nervous system, blood, kidneys, intestinal tract) through prolonged or repeated exposure. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear eye protection, protective gloves, protective clothing. Wash exposed skin thoroughly after handling. Avoid release to the environment. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |