missing translation for 'onlineSavingsMsg'
Learn More

Mercury AA Standard, 1000 ppm Hg in 3% HNO3, Ricca Chemical

Catalog No. AHG1KN500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AHG1KN500 500 mL Natural Poly Bottle
AHG1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. AHG1KN500 Supplier Ricca Chemical Company Supplier No. AHG1KN500
Only null left
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 21908-53-2 , 7697-37-2 , 7732-18-5
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Hg
CAS Min % 93.0, 4.63, 0.1
CAS Max % 97.0, 4.81, 0.1
Equivalence 1 mL = 1 mg Hg
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Quantity 500 mL
Chemical Name or Material Mercury AA Standard
Show More Show Less