missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury AA Standard, 1000 ppm Hg in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 1,000ppm Hg |
| CAS Min % | 93.0, 4.63, 0.1 |
| CAS Max % | 97.0, 4.81, 0.1 |
| Equivalence | 1 mL = 1 mg Hg |
| Physical Form | Liquid |
| Color | Colorless |
| pH | < 2 |
| Melting Point | Approximately 0°C |
| Quantity | 100 mL |
| Show More |