missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Chloride TS, Ricca Chemical

Catalog No. 466016
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
466016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. 466016 Supplier Ricca Chemical Company Supplier No. 466016
Only null left
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 7487-94-7 , 7732-18-5
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name mercury(2+) dichloride
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Color Colorless
CAS Min % 92.0, 5.98
CAS Max % 96.0, 6.23
Grade Laboratory
Packaging Natural Poly Bottle
pH 3.2
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Mercuric Chloride, Test Solution