missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Acetate, 25 g/L in Glacial Acetic Acid, Ricca Chemical

Catalog No. R4625000500
Change view
Click to view available options
Package: Safety-Coated Glass; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R4625000500 Package: Safety-Coated Glass; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4625000500 Supplier Ricca Chemical Company Supplier No. R4625000500C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1600-27-7, 64-19-7
CAS Min % 96.0, 2.24
CAS Max % 100.0, 2.33
Molecular Formula C4H6HgO4
Synonym mercury 2+ ion acetic acid
InChI Key BRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].[Hg+2]
IUPAC Name mercury(2+);diacetate
Molecular Weight (g/mol) 318.68
PubChem CID 15337
ChEBI CHEBI:33211
Packaging Safety-coated Glass Bottle
Solvent or Matrix Glacial Acetic Acid
Quantity 500 mL
Concentration or Composition Notes 25g/L
Chemical Name or Material Mercuric Acetate
Show More Show Less