missing translation for 'onlineSavingsMsg'
Learn More

Mercaptosuccinic acid, 98%

Catalog No. AAB2330114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2330114 25 g
AAB2330122 100 g
AAB2330136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2330114 Supplier Thermo Scientific Chemicals Supplier No. B2330114

CAS: 70-49-5 | C4H6O4S | 150.148 g/mol

Mercaptosuccinic acid is used in the preparation of mercaptosuccinic acid diethyl ester by reaction with ethanol. It is also used as a brightening agent in metal plating. Further, it serves as an intermediate for the synthesis of corrosion inhibitors, soil fumigants, active pharmaceutical ingredients and electroplating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercaptosuccinic acid is used in the preparation of mercaptosuccinic acid diethyl ester by reaction with ethanol. It is also used as a brightening agent in metal plating. Further, it serves as an intermediate for the synthesis of corrosion inhibitors, soil fumigants, active pharmaceutical ingredients and electroplating agents.

Solubility
Soluble in water, alcohol and acetone. Slightly soluble in ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 70-49-5
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004860
InChI Key NJRXVEJTAYWCQJ-UHFFFAOYSA-N
Synonym mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid
PubChem CID 6268
ChEBI CHEBI:38705
IUPAC Name 2-sulfanylbutanedioic acid
SMILES C(C(C(=O)O)S)C(=O)O

Specifications

Melting Point 150°C to 155°C
Quantity 25 g
Beilstein 1099858
Merck Index 14,9343
Solubility Information Soluble in water,alcohol and acetone. Slightly soluble in ether. Insoluble in benzene.
Formula Weight 150.15
Percent Purity 98%
Chemical Name or Material Mercaptosuccinic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-736-4
RTECSNumber WM8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only