missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mequitazine

Supplier:  Thermo Scientific Chemicals 466930050

 View more versions of this product

Catalog No. AC466930050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Mequitazine
5 g
148.0°C
322.47
C(C1CN2CCC1CC2)N1C2=CC=CC=C2SC2=CC=CC=C12
322.47
29216-28-2
Solubility aq. soln.: insoluble. Other solubilities: soluble in methanol, ethanol
C20H22N2S
HOKDBMAJZXIPGC-UHFFFAOYNA-N
10-({1-azabicyclo[2.2.2]octan-3-yl}methyl)-10H-phenothiazine

Chemical Identifiers

29216-28-2
C20H22N2S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

GHS Signal Word: Danger

EINECSNumber : 249-521-7

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only