accessibility menu, dialog, popup

UserName

NNS

CAS RN 29216-28-2

IUPAC Name: 10-({1-azabicyclo[2.2.2]octan-3-yl}methyl)-10H-phenothiazine
Synonyms: mequitazine primalan quitadrill vigigan instotal kitazemin mircol zesulan virginan metaplexan
Molecular Weight (g/mol): 322.47
Molecular Formula: C20H22N2S
InChi Key: HOKDBMAJZXIPGC-UHFFFAOYNA-N
SMILES: C(C1CN2CCC1CC2)N1C2=CC=CC=C2SC2=CC=CC=C12
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results