missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mequitazine

Catalog No. AC466930010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466930010 1 g
AC466930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466930010 Supplier Thermo Scientific Chemicals Supplier No. 466930010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29216-28-2
Molecular Formula C20H22N2S

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: insoluble. Other solubilities: soluble in methanol, ethanol
Melting Point 148.0°C
Formula Weight 322.47
InChI Key HOKDBMAJZXIPGC-UHFFFAOYNA-N
SMILES C(C1CN2CCC1CC2)N1C2=CC=CC=C2SC2=CC=CC=C12
IUPAC Name 10-({1-azabicyclo[2.2.2]octan-3-yl}methyl)-10H-phenothiazine
Molecular Weight (g/mol) 322.47
Chemical Name or Material Mequitazine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

EINECSNumber 249-521-7

RUO – Research Use Only