missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Menthone 90.0+%, TCI America™
$79.74 - $645.52
Chemical Identifiers
| CAS | 14073-97-3 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.25 |
| MDL Number | MFCD00001634,MFCD00136033 |
| InChI Key | NFLGAXVYCFJBMK-BDAKNGLRSA-N |
| Synonym | --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one |
| PubChem CID | 26447 |
| ChEBI | CHEBI:15410 |
| IUPAC Name | (2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one |
| SMILES | CC(C)[C@@H]1CC[C@@H](C)CC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
M051325ML
|
N/A | 25 mL |
Each for $79.74
|
|
|||||
|
M0513100ML
|
N/A | 100 mL |
Each for $203.15
|
|
|||||
|
M0513500ML
|
N/A | 500 mL |
Each for $645.52
|
|
|||||
Chemical Identifiers
| 14073-97-3 | |
| 154.25 | |
| NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
| 26447 | |
| (2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one |
| C10H18O | |
| MFCD00001634,MFCD00136033 | |
| --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
| CHEBI:15410 | |
| CC(C)[C@@H]1CC[C@@H](C)CC1=O |
Specifications
| 14073-97-3 | |
| 88°C | |
| MFCD00001634,MFCD00136033 | |
| NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
| (2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one | |
| 26447 | |
| 154.25 | |
| Liquid |
| Yellow | |
| C10H18O | |
| --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
| CC(C)[C@@H]1CC[C@@H](C)CC1=O | |
| 154.25 | |
| CHEBI:15410 | |
| ≥90.0% (GC) | |
| (-)-Menthone |
Safety and Handling
EINECSNumber : (3)-2398
TSCA : Yes