missing translation for 'onlineSavingsMsg'
Learn More

(-)-Menthone 90.0+%, TCI America™

Catalog No. M051325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M051325ML 25 mL
M0513100ML 100 mL
M0513500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M051325ML Supplier TCI America Supplier No. M051325ML

Chemical Identifiers

CAS 14073-97-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001634,MFCD00136033
InChI Key NFLGAXVYCFJBMK-BDAKNGLRSA-N
Synonym --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
PubChem CID 26447
ChEBI CHEBI:15410
IUPAC Name (2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one
SMILES CC(C)[C@@H]1CC[C@@H](C)CC1=O

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Menthone
EINECSNumber (3)-2398
TSCA Yes