missing translation for 'onlineSavingsMsg'
Learn More

(-)-Menthone 90.0+%, TCI America™

Supplier:  TCI America M0513100ML

Quantity 100 mL

 View more versions of this product

Catalog No. M0513100ML


Only null left

Description

Description

Specifications

Chemical Identifiers

14073-97-3
154.25
NFLGAXVYCFJBMK-BDAKNGLRSA-N
26447
(2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one
C10H18O
MFCD00001634,MFCD00136033
--menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
CHEBI:15410
CC(C)[C@@H]1CC[C@@H](C)CC1=O

Specifications

(-)-Menthone
Yellow
C10H18O
MFCD00001634,MFCD00136033
NFLGAXVYCFJBMK-BDAKNGLRSA-N
(2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-one
26447
154.25
Liquid
14073-97-3
88°C
100 mL
--menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
CC(C)[C@@H]1CC[C@@H](C)CC1=O
154.25
CHEBI:15410
≥90.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2398

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions