missing translation for 'onlineSavingsMsg'
Learn More

Menthone, ≥97% (Mixture of Isomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111019867
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019867 1 mL
111018394 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019867 Supplier Merck Emd Millipore Supplier No. 954011ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 10458-14-7
Density 0.896 g/mL (at 20°C)
Flash Point 69°C
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00062998
Boiling Point 85°C to 88°C (12 mmHg)
Synonym 2-Isopropyl-5-methylcyclohexanone
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥97% (mixture of isomers, GC)
Grade Analytical Standard
Show More Show Less