missing translation for 'onlineSavingsMsg'
Learn More

Menthofuran, 95%

Catalog No. AC413560500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413560500 50 g Glass bottle
AC413560100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413560500 Supplier Thermo Scientific Chemicals Supplier No. 413560500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 494-90-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 494-90-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00041851
InChI Key YGWKXXYGDYYFJU-UHFFFAOYSA-N
Synonym menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan
PubChem CID 329983
ChEBI CHEBI:50542
IUPAC Name 3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
SMILES CC1CCC2=C(C1)OC=C2C

Specifications

Color Yellow
Density 0.9700g/mL
Boiling Point 87.0°C (15.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4832 to 1.4852
Quantity 50 g
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Formula Weight 150.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Menthofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 207-795-5

RUO – Research Use Only