missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menaquinone (K2), MilliporeSigma™ Supelco™
Description
Vitamines pharmaceutiques (petites molécules), nutraceutiques et produits naturels
Spécifications
Spécifications
| Packaging | Ampoule |
| CAS | 863-61-6 |
| Physical Form | Sympa |
| Linear Formula | C31H40O2 |
| MDL Number | MFCD00079646 |
| Synonym | Vitamin K2; Menaquinone 4; Menatetrenone |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 444.65 |
| Grade | Norme analytique |