missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Flame Resistant Mock Turtleneck
Men's Lightweight Navy Flame Resistant Mock Turtleneck is a comfortable and durable CAT 2 AR/FR, NFPA™ 2112 compliant mock turtleneck that will keep you warm and protected on the job.
$142.86 - $142.86
Specifications
| Gender | Men's |
|---|---|
| Material | FR Cotton |
| Color | Navy |
| Neck Style | Collar |
| Sleeve Style | Long Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-028
|
Bulwark™
SEK2NVRGS |
Small |
Each for $142.86
|
|
|||||
|
17-204-029
|
Bulwark™
SEK2NVRGM |
Medium |
Each for $142.86
|
|
|||||
|
17-204-129
|
Bulwark™
SEK2NVRGL |
Large |
Each for $142.86
|
|
|||||
|
17-204-021
|
Bulwark™
SEK2NVRGXL |
X-Large |
Each for $142.86
|
|
|||||
|
17-204-148
|
Bulwark™
SEK2NVRGXXL |
2X-Large |
Each for $142.86
|
|
|||||
|
17-204-367
|
Bulwark™
SEK2NVRG3XL |
3X-Large |
Each for $142.86
|
|
|||||
|
17-204-030
|
Bulwark™
SEK2NVRG4XL |
4X-Large |
Each for $142.86
|
|
|||||
|
17-204-127
|
Bulwark™
SEK2NVLNL |
Large (Long) |
Each for $142.86
|
|
|||||
|
17-204-150
|
Bulwark™
SEK2NVLNXL |
X-Large (Long) |
Each for $142.86
|
|
|||||
|
17-204-128
|
Bulwark™
SEK2NVLNXXL |
2X-Large (Long) |
Each for $142.86
|
|
|||||
Description
- A comfortable and durable CAT 2 AR/FR, NFPA™ 2112 compliant mock turtleneck that will keep you warm and protected on the job.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Pull On
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (198.4 g/m&superscript_2;) Interlock
- Blend: 100% Cotton
- Collar: Double-layer, rib knit collar sized to protect the neck
- Features: Double-layer, rib knit collar sized to protect the neck, Tagless brand label and warning label
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 8.9
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Unisex
- Materials: Cotton
- Brand: Bulwark™
Specifications
| Men's | |
| Navy | |
| Long Sleeve | |
| Reusable | |
| 8.9 |
| FR Cotton | |
| Collar | |
| Pull On | |
| Category 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |