missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Navy Flame Resistant Mock Turtleneck

Catalog No. 17204028
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204028 Small
17204029 Medium
17204129 Large
17204021 X-Large
17204148 2X-Large
17204367 3X-Large
17204030 4X-Large
17204127 Large (Long)
17204150 X-Large (Long)
17204128 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17204028 Supplier Bulwark™ Supplier No. SEK2NV RG S

Men's Lightweight Navy Flame Resistant Mock Turtleneck is a comfortable and durable CAT 2 AR/FR, NFPA™ 2112 compliant mock turtleneck that will keep you warm and protected on the job.

  • A comfortable and durable CAT 2 AR/FR, NFPA™ 2112 compliant mock turtleneck that will keep you warm and protected on the job.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Pull On
  • Wash Care: Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 7 oz. (198.4 g/m&superscript_2;) Interlock
  • Blend: 100% Cotton
  • Collar: Double-layer, rib knit collar sized to protect the neck
  • Features: Double-layer, rib knit collar sized to protect the neck, Tagless brand label and warning label
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 8.9
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Unisex
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size Small
Gender Men's
Material FR Cotton
Color Navy
Neck Style Collar
Sleeve Style Long Sleeve
Closure Type Pull On
Disposable Reusable
No. of Pockets None
Hazard Risk Category Category 2
Arc Flash Rating 8.9
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less