missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Flame Resistant Mock Turtleneck
Men's Lightweight Navy Flame Resistant Mock Turtleneck is a comfortable and durable CAT 2 AR/FR, NFPA™ 2112 compliant mock turtleneck that will keep you warm and protected on the job.
Supplier: Bulwark™ SEK2NVLNXXL
Description
- A comfortable and durable CAT 2 AR/FR, NFPA™ 2112 compliant mock turtleneck that will keep you warm and protected on the job.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Pull On
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (198.4 g/m&superscript_2;) Interlock
- Blend: 100% Cotton
- Collar: Double-layer, rib knit collar sized to protect the neck
- Features: Double-layer, rib knit collar sized to protect the neck, Tagless brand label and warning label
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 8.9
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Unisex
- Materials: Cotton
- Brand: Bulwark™
Specifications
| 2X-Large (Long) | |
| FR Cotton | |
| Collar | |
| Pull On | |
| None | |
| 8.9 |
| Men's | |
| Navy | |
| Long Sleeve | |
| Reusable | |
| Category 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |