Learn More
Memmert Usa Llc Memmert Universal Heating and Drying Laboratory Oven UF160Plus. 160L - Programmable Model - 230 Volt, Forced-Air Convection, Stainless Steel Construction with TwinDISPLAY digital controller.
Supplier: Memmert UF160PLUS230V
Memmert Universal heating and drying Oven - this programmable model is the 230 volt, forced-air convection, with TwinDISPLAY digital controller. Interior dimensions 560/720/361-W/H/D mm. Includes (2) Stainless Steel Grid Shelves, Atmocontrol Software, Atmocontrol Manual, Operation manual, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.