Chemglass Inc Melting Point Tubes, Closed One End, 1.5mm-1.8mm O.D. x 0.20mm Wall, Vial of 100, 90mm Length
Numéro de catalogue. CG184101
Melting Point Tubes, Closed One End, 1.5mm-1.8mm O.D. x 0.20mm Wall, Vial of 100, 90mm Length. Similar to CG-1840, but closed at one end. Tube is manufactured from borosilicate glass and packaged 100 tubes per vial. The Code -02 has a smaller I.D. to conform to U.S.P XXI requirements.
Numéro de catalogue.
CG184101
Fournisseur
Chemglass IncCode fournisseur
CG184101
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Melting Point Tubes, Closed One End, 1.5mm-1.8mm O.D. x 0.20mm Wall, Vial of 100, 90mm Length. Similar to CG-1840, but closed at one end. Tube is manufactured from borosilicate glass and packaged 100 tubes per vial. The Code -02 has a smaller I.D. to conform to U.S.P XXI requirements.