missing translation for 'onlineSavingsMsg'
Learn More

Melting point Standard 79-81C, MilliporeSigma™ Supelco™

Catalog No. 111020159
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020159 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020159 Supplier Merck Emd Millipore Supplier No. 014221G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Melting point standard 79-81C (Naphthalene) is a hydrocarbon which is nonpolar in nature. It is soluble in toluene. It can be crystallized from ether.

Specifications

CAS 91-20-3
Ignition Point 978°F
Vapor Pressure 0.03 mmHg (25°C)
Vapor Density 4.4 (vs. air)
Flash Point 80°C
Linear Formula C10H8
MDL Number MFCD00001742
Melting Point 79°C to 81°C (±0.3 C); 80°C to 82°C
Boiling Point 218°C
Synonym Naphthalene
Type Analytical Standard
Quantity 1 g
Formula Weight 128.17
Grade Analytical Standard
Show More Show Less