missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Melatonin
Description
Endogenous hormone that acts as an agonist at melatonin receptors MT1 and MT2. Exhibits a prominent role in the control of circadian rhythm, displays immunomodulatory activity and acts as a powerful antioxidant in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 232.28 |
| Purity | >99% |
| Molecular Weight (g/mol) | 232.283 |
| Chemical Name or Material | Melatonin |
Safety and Handling
| Recommended Storage | Store at -20°C |