Learn More
Integra™ Miltex™ MeisterHand™ Scalpel Handles
Catalog No. 12460635
Change view
Click to view available options
Size:
# 3
# 3L
# 4
Length (English):
5 in.
5.25 in.
8.25 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English)
12460635 # 4 5.25 in.
12460633 # 3L 8.25 in.
12460634 # 3 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12460635 Supplier Integra™ Miltex™ Supplier No. MH48
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
  • Extra fine
  • Stainless steel
  • With mm and cm graduations on No. 3 and No. 4
  • Non-glare mattefinish
Specifications
Blade Material Stainless Steel
Product Type Scalpel Handle
Size # 4
Length (English) 5.25 in.
Length (Metric) 13.5 cm
For Use With (Equipment) # 20 to # 25 Blade for Deep Surgery
Description Non-glare Matte
Graduated Yes