missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medroxyprogesterone Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Ampule |
| CAS | 71-58-9 |
| Linear Formula | C24H34O4 |
| MDL Number | MFCD00010483 |
| Melting Point | 206°C to 207°C |
| Synonym | Medroxyprogesterone 17-acetate; 17 alpha-Acetoxy-6 alpha-methylprogesterone; 17 alpha-Hydroxy-6 alpha-methyl-4-pregnene-3,20-dione 17-acetate; 6 alpha-Methyl-17 alpha-acetoxyprogesterone; 6 alpha-Methyl-17 alpha-hydroxyprogesterone acetate |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 386.52 |
| Grade | Certified Reference Material |